Learn More
Medchemexpress LLC N-(cis-4-{[4-(dimethylamino)quinazolin-2-yl]amino}cyclohexyl)-3,4-difluorobenzamide HCl | 510733-97-8 | MFCD08276913 | 99.8% | 461.94 g/mol | C23H26ClF2N5O | 100 MG

Supplier: Medchemexpress LLC HY107624100MG
ATC0175 is a small-molecule, selective melanin-concentrating hormone receptor 1 (MCH1R) antagonist intended for preclinical research in neuropharmacology. It is supplied as a high-purity solid for use in in vitro and in vivo pharmacology studies.
- Selective MCH1R antagonist suitable for pharmacology research.
- High purity (99.8% by HPLC).
- Molecular weight 461.94 g/mol and formula C23H26ClF2N5O.
- CAS number 510733-97-8 for unambiguous identification.
- Supplied as a 100 MG solid appropriate for formulation and dosing.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.