Learn More
Medchemexpress LLC N-cyclohexyl-2-benzothiazolesulfenamide | 95-33-0 | MFCD00022872 | 100.0% | 264.42 g·mol⁻¹ | C13H16N2S2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02075510G
Encompass_Preferred
Thiohexam (N-cyclohexyl-2-benzothiazolesulfenamide; CAS 95-33-0) is a sulfur-containing accelerator commonly used in rubber vulcanization and supplied as an analytical/research standard. The compound has formula C13H16N2S2 and a molecular weight of approximately 264.42 g·mol⁻¹. It is a known dermatological sensitizer and allergen; consult the SDS for hazard, handling, and storage information.
  • Used as a rubber cure accelerator.
  • Suitable for analytical and research applications.
  • High purity supports analytical assays.
  • Sulfur-containing benzothiazole chemical structure.
  • Known dermatological sensitizer; requires appropriate personal protective equipment.
Catalog No. 50-003-01359