Learn More
Medchemexpress LLC N'-cyclohexyl-N-(phenylmethyl)-N-(4-piperidinylmethyl)-urea | 1629138-41-5 | 99.6% | 329.48 g/mol | C20H31N3O | 200 MG
SDP
Supplier:  Medchemexpress LLC HY100347200MG
Encompass_Preferred
SRI-011381 is an orally active transforming growth factor-β (TGF-β)/Smad signaling agonist reported to exhibit neuroprotective effects. Intended for research use only, the compound is provided with a defined chemical formula, CAS identifier, and analytical data to support laboratory studies.
  • Orally active TGF-β signaling agonist suitable for in vitro and in vivo studies.
  • Reported neuroprotective effects in preclinical models.
  • High purity analytical grade for consistent experimental results.
  • Available as solid and as solution in DMSO for flexible use.
  • Characterized by a defined molecular weight for dosing calculations.
Catalog No. 50-002-14135