Learn More
Medchemexpress LLC N-cyclohexylmaleimide | 1631-25-0 | MFCD00043904 | 99.5% | 179.22 g·mol⁻¹ | C10H13NO2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW016550100G
Encompass_Preferred
N-cyclohexylmaleimide is a maleimide-containing organic reagent (CAS 1631-25-0) used in chemical synthesis and bioconjugation. The maleimide functional group reacts selectively with thiols and can participate in reactions with NHS esters, making the compound useful for crosslinking, probe derivatization, and polymer functionalization.
  • Reacts selectively with thiols under mild conditions.
  • Enables conjugation to proteins and peptides.
  • Useful for crosslinking and polymer functionalization.
  • Available as a solid with high reported purity suitable for synthesis.
  • Compatible with common organic solvents and standard lab conditions.
Catalog No. 50-003-01142