Learn More
Medchemexpress LLC N-cyclohexylmaleimide | 1631-25-0 | MFCD00043904 | 99.5% | 179.22 g·mol⁻¹ | C10H13NO2 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW016550
Encompass_Preferred
N-cyclohexylmaleimide is a maleimide-containing reagent that selectively reacts with thiol groups for bioconjugation and chemical modification. Supplied as a high-purity solid, it is commonly used in peptide modification, crosslinking, and the synthesis of functionalized small molecules where selective thiol labeling is required.
  • Reacts selectively with thiol groups.
  • Suitable for bioconjugation and peptide modification.
  • High purity (99.48%).
  • Supplied as a solid with defined storage recommendations.
  • Available in multiple pack sizes for scale flexibility.
Catalog No. 50-003-02316