Learn More
Medchemexpress LLC N-cyclopropyl-2,4-difluoro-5-((2-(pyridin-2-ylamino)thiazol-5-ylmethyl)amino)benzamide | 639858-32-5 | 99.6% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY120640100MG
Encompass_Preferred
BMS-605541 is a selective, orally active VEGFR-2 kinase inhibitor used in cancer research. It demonstrates potent inhibition of VEGFR-2 and measurable activity against VEGFR-1 and PDGFR-β.
  • Selective VEGFR-2 (Flk-1) inhibition with low nanomolar potency.
  • Demonstrated activity against VEGFR-1 and PDGFR-β for pathway studies.
  • High purity (99.6%) suitable for in vitro and cellular research.
  • Available in multiple solid pack sizes and as a 10 mM solution in DMSO.
  • Commonly used in cancer research and VEGF/PDGF signaling studies.
Catalog No. 50-002-33842