Learn More
Medchemexpress LLC N-DBCO-N-bis(PEG2-C2-acid) | 2110449-00-6 | 96.7% | C33H40N2O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14054350MG
Encompass_Preferred
N-DBCO-N-bis(PEG2-C2-acid) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It also functions as a click chemistry reagent, containing a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs are composed of two different ligands connected by a linker, enabling selective degradation of target proteins via the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent with a DBCO group
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables selective degradation of target proteins
Catalog No. 50-003-56932