Learn More
Medchemexpress LLC N-deacetyl-N-formylcolchicine | 7411-12-3 | 385.41 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYZ8644
Encompass_Preferred
N-Deacetyl-N-formylcolchicine is a selective inhibitor targeting the colchicine-binding site of tubulin. It demonstrates antiproliferative activity against various cancer cell lines. This compound inhibits microtubule polymerization, blocking cell division, inducing cancer cell apoptosis, and suppressing migration, showing promise for research in cancers like lung cancer and various solid tumors.
  • Selective inhibitor targeting the colchicine-binding site of tubulin
  • Demonstrates antiproliferative activity against various cancer cell lines
  • Inhibits microtubule polymerization
  • Blocks cell division
  • Induces cancer cell apoptosis
  • Suppressing cell migration
  • Shows promise for research in lung cancer and various solid tumors
Catalog No. 50-003-65700