Learn More
Medchemexpress LLC N-decyl-N,N-dimethyldecan-1-aminium chloride | 7173-51-5 | MFCD00066262 | 98.0% | 362.08 g/mol | C22H48ClN | 1 ML

Supplier: Medchemexpress LLC HYW04218110MM/1ML
N-Decyl-N,N-dimethyldecan-1-aminium chloride is a quaternary ammonium compound supplied as a small-volume solution for laboratory use. It is provided as a 10 mM solution in DMSO and is intended for use as a reagent or standard in biochemical assays and biocide-related research.
- 10 mM solution in DMSO, 1 mL total volume.
- Purity approximately 98.0% as specified on the certificate of analysis.
- Chemical formula: C22H48ClN; molecular weight: 362.08 g/mol.
- CAS number 7173-51-5 for unambiguous identification and safety referencing.
- Suitable for assay development, compatibility testing, and use as a reagent standard.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.