Learn More
Medchemexpress LLC N-Demethylricinine | 21642-98-8 | 99.94% | C7H6N2O2 | 100 G

Supplier: Medchemexpress LLC HY65008100G
N-Demethylricinine is a ricinine that can be interconverted with ricinine in senescent and green castor plant leaves. Ricinine is an α-pyridone alkaloid biosyntynthetically related to the pyridine nucleotide cycle. The alkaloid catabolism of ricinine is associated with the aging process. It is intended for research use only.
- Appearance: Solid
- Color: Off-white to yellow
- Molecular weight: 150.13
- Structure classification: Alkaloids
- Initial source: Plants Euphorbiaceae Ricinus communis L.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.