Learn More
Medchemexpress LLC N-desalkylflurazepam | 2886-65-9 | MFCD00083443 | 100.0% | 288.70 g/mol | C15H10ClFN2O | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1064545MG
Encompass_Preferred
N-desalkylflurazepam is a benzodiazepine metabolite supplied as a high-purity analytical standard for research and analytical applications. It is suited for chromatographic and mass spectrometric assays, method development, and quality control.
  • Analytical standard for benzodiazepine metabolite testing.
  • High purity for reliable reference and quality-control measurements.
  • Molecular formula C15H10ClFN2O and molecular weight 288.70 g/mol.
  • Available in small milligram pack sizes tailored for analytical use.
  • Provided with datasheet detailing handling and storage.
Catalog No. 50-002-40988