Learn More
Medchemexpress LLC N-desalkylflurazepam | 2886-65-9 | MFCD00083443 | 100.0% | 288.70 g/mol | C15H10ClFN2O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10645450MG
Encompass_Preferred
N-desalkylflurazepam is a benzodiazepine metabolite provided as a high-purity research standard for analytical, pharmacokinetic, and metabolic studies. The compound has molecular formula C15H10ClFN2O and a molecular weight of 288.70 g/mol, and is supplied in small-mass quantities intended for laboratory use.
  • High purity suitable for analytical and pharmacokinetic assays.
  • Available in small pack sizes with a 50 mg option for bench-scale work.
  • Clearly identified by CAS number 2886-65-9 for unambiguous reference.
  • Appropriate as a reference standard in metabolic, toxicological, and method development studies.
  • Molecular formula C15H10ClFN2O and molecular weight 288.70 g/mol documented for accurate calculations.
Catalog No. 50-002-40037