Learn More
Medchemexpress LLC N-desalkylflurazepam | 2886-65-9 | MFCD00083443 | 98.0% | 288.70 g/mol | C15H10ClFN2O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY106454100MG
Encompass_Preferred
N-desalkylflurazepam is an analytical standard of a long-acting benzodiazepine metabolite (norfludiazepam) provided for research and analytical applications. It is intended for use as a reference material in method development, quantitative assays, and pharmacology studies requiring a characterized metabolite standard.
  • Analytical standard of a benzodiazepine metabolite for reference and calibration.
  • High purity suitable for analytical assays (98.0%).
  • Chemical formula C15H10ClFN2O and molecular weight 288.70 g/mol.
  • Appropriate for LC-MS, HPLC, and quantitative method development.
  • Supplied as a characterized solid for research use.
Catalog No. 50-002-41027