Learn More
Medchemexpress LLC N-Descyclopropanecarbaldehyde Olaparib | 763111-47-3 | 99.46% | C20H19FN4O2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY75706
Encompass_Preferred
N-Descyclopropanecarbaldehyde Olaparib is an analogue of Olaparib containing a DOTA moiety and acts as a PARP inhibitor. It is used for synthesizing novel dual EGFR and PARP PROTAC, DP-C-4. This compound can be radiolabeled with F-18 or a fluorophore for positron emission tomography (PET) or optical imaging in various tumor types.
  • Analogue of Olaparib containing a DOTA moiety
  • Functions as a PARP inhibitor
  • Used for synthesizing novel dual EGFR and PARP PROTAC, DP-C-4
  • Can be radiolabeled with F-18 or a fluorophore for PET or optical imaging
  • Intended solely for research purposes
Catalog No. 50-003-58617