Learn More
Medchemexpress LLC N-Desethyl amodiaquine-d5 | 1173023-19-2 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY128554S1MG
Encompass_Preferred
N-Desethyl amodiaquine-d5 is the deuterium labeled N-Desethyl amodiaquine, which is the major biologically active metabolite of Amodiaquine. It is an antiparasitic agent with IC50 values for strains V1/S and 3D7 at 97 nM and 25 nM, respectively.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-41877