Learn More
Medchemexpress LLC N-Desethyl amodiaquine dihydrochloride | 79049-30-2 | 99.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY128554A1MG
Encompass_Preferred
N-Desethyl amodiaquine dihydrochloride is the biologically active metabolite of Amodiaquine. It functions as an antiparasitic agent, demonstrating inhibitory effects on _Plasmodium falciparum_ strains V1/S and 3D7 with IC50 values of 97 nM and 25 nM, respectively. This compound is suitable for malaria research.
  • Biologically active metabolite of Amodiaquine
  • Functions as an antiparasitic agent
  • Demonstrates inhibitory effects on _Plasmodium falciparum_ strains V1/S and 3D7
  • Suitable for malaria research
Catalog No. 50-003-41945