Learn More
Cayman Chemical N-desethyl SunItInIb 500ug
SDP
Supplier:  Cayman Chemical 22292500 UG
Encompass_Preferred
An active metabolite of sunitinib; formed when sunitinib undergoes N-de-ethylation by CYP3A4; is pharmacologically active having similar inhibitory activity to sunitinib
Catalog No. 50-298-7657