Learn More
Cayman Chemical N-Desethyl VardenafIl 500UG
SDP
Supplier:  Cayman Chemical 9001800500 UG
Encompass_Preferred
A metabolite of vardenafil that is formed by the action of CYP3A4 and CYP3A5
Catalog No. 50-289-3725