Learn More
TARGETMOL CHEMICALS INC N-desmethyl Enzalutamide 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1945025MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. N-desmethyl Enzalutamide (N-desmethyl MDV 3100) is the active Enzalutamide metabolite.It is the active metabolite of Enzalutamide. N-desmethyl Enzalutamide demonstrates primary and secondary pharmacodynamics of similar potency to Enzalutamide and circulates at approximately the same plasma concentrations as enzalutamide. Purity 99.74%
Catalog No. 50-287-6015