Learn More
Medchemexpress LLC N-desmethyl imatinib | 404844-02-6 | MFCD07369291 | 98.3% | 479.59 | C28H29N7O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYG001710MG
Encompass_Preferred
N-Desmethyl imatinib (norimatinib) is the primary N-desmethyl metabolite of imatinib and acts as a c-Abl tyrosine kinase inhibitor. It is supplied as a research reagent for biochemical and cellular studies, including kinase inhibition and neurodegenerative disease models.
  • High purity suitable for analytical and biological assays (purity 98.3%).
  • Available as a small solid quantity and as a ready-to-use DMSO solution for reconstitution.
  • Characterized molecular weight and formula for accurate dosing and calculations.
  • Useful in kinase inhibition and neurodegeneration research applications.
  • Provided with supporting datasheet and safety data sheet for laboratory handling.
Catalog No. 50-002-96991