Learn More
TARGETMOL CHEMICALS INC N-Desmethyl imatinib 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T116415MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes.N-Desmethyl imatinib (Imatinib metabolite N-Desmethyl imatinib) is a metabolite of Imatinib, which is a multi-target inhibitor of c-Kit, v-Abl, and PDGFR. Purity 98.6%
Catalog No. 50-287-9487