Learn More
Selleck Chemical LLC N-Desmethyl tamoxifen HCl E2368-5MG
SDP
Supplier:  Selleck Chemical LLC E23685MG
Encompass_Preferred
N-Desmethyltamoxifen hydrochloride (N-Desmethyl tamoxifen HCl) the primary metabolite of tamoxifen is an estrogen response modifer and protein kinase C inhibitor with IC50 of 25 M
Catalog No. 50-000-40684