Learn More
Medchemexpress LLC N-desmethyl venlafaxine hydrochloride | 93413-90-2 | MFCD18711349 | 99.9% | C16H26ClNO2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY131527R10MG
Encompass_Preferred
Wy-45494 hydrochloride (N-desmethyl venlafaxine hydrochloride) is the hydrochloride salt of a minor active metabolite of the SNRI venlafaxine. It is supplied as an analytical standard for research, quality control, and method development in chromatographic and mass spectrometry applications.
  • High purity: 99.85% (analytical standard).
  • Suitable for HPLC, GC, and MS method development and quantitation.
  • Hydrochloride salt form provides enhanced solubility and stability.
  • Chemical formula C16H26ClNO2 and molecular weight ~299.84 g/mol.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-61967