Learn More
Medchemexpress LLC HY-129099A 5mg Medchemexpress, N-Desmethyltamoxifen (hydrochloride) CAS:15917-65-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY129099A5MG
Encompass_Preferred
Medchemexpress, HY-129099A 5mg N-Desmethyltamoxifen (HCl) CAS:15917-65-4 N-Desmethyltamoxifen hydrochloride is the major metabolite of tamoxifen in humans. N-Desmethyltamoxifen, a poor antiestrogen, is a ten-fold more potent protein kinase C (PKC) inhibitor than Tamoxifen. N-Desmethyltamoxifen hydrochloride is also a potent regulator of ceramide metabolism in human AML cells, limiting ceramide glycosylation, hydrolysis, and sphingosine phosphorylation[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9795