Learn More
Cayman Chemical N-Despropyl RopInIrole 50mg
SDP
Supplier:  Cayman Chemical 3415650 MG
Encompass_Preferred
An active metabolite of ropinirole; increases extracellular acidification rates in CHO cells expressing human dopamine D2, D3, and D4 receptors (EC50s = 0.63, 0.063, and 1.23 µM, respectively); a potential impurity found in commercial preparations of ropinirole
Catalog No. 50-294-8302