Learn More
Medchemexpress LLC N-(diphenylmethylene) glycine benzyl ester | 81477-91-0 | MFCD08062206 | 98.0% | 329.40 g·mol-1 | C22H19NO2
SDP
Supplier:  Medchemexpress LLC HYW0186205G
Encompass_Preferred
Diphenylmethylene-glycine benzyl ester is a protected glycine derivative used as a synthetic intermediate and research reagent. The compound is the benzyl ester of glycine bearing a diphenylmethylene (benzhydrylidene) protecting group; it is supplied as a white to off-white solid with high purity for organic synthesis and peptide chemistry.
  • Protected glycine derivative suitable for peptide and small-molecule synthesis.
  • Benzyl ester functionality allows straightforward hydrogenolytic deprotection.
  • Diphenylmethylene group protects the amino function during reactions.
  • High purity (~98.0%) appropriate for research applications.
  • Multiple pack sizes available to support different scales.
  • Stable when stored under recommended conditions for extended shelf life.
Catalog No. 50-003-01023