Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(diphenylmethylene) glycine benzyl ester | 81477-91-0 | MFCD08062206 | ≥98.0% | 329.40 g/mol | C22H19NO2 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW01862010G

Encompass_Preferred

N-(diphenylmethylene) glycine benzyl ester is an N-protected glycine benzyl ester used as a synthetic intermediate in amino-acid and peptide chemistry. It is supplied as a solid laboratory reagent suitable for organic synthesis and research applications.

  • Protected glycine derivative useful for peptide coupling and amino-acid modification.
  • White to off-white solid powder.
  • Molecular formula C22H19NO2; molecular weight 329.40 g/mol.
  • Purity ≥98% as reported by suppliers.
  • Store powder at -20°C for long term; store solutions at -80°C for extended stability.
  • Available in multiple laboratory pack sizes.

Catalog No. 50-003-02009


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.