Learn More
Research Products International Corp n-Dodecyl-B-D-maltoside, 10 G
SDP
Supplier:  Research Products International Corp D1200010.0
Encompass_Preferred
Non-ionic detergent used in membrane protein studies. Reported to increase thermal stability of lipid-free rhodopsin in solution.
Catalog No. 50-197-7073