Learn More
AdipoGen n-Dodecyl-beta-D-maltoside
SDP
Supplier:  AdipoGen AGCC10010G001
Encompass_Preferred
Chemical. CAS 69227-93-6. Formula C24H46O11. MW 510.6. Water-soluble non-ionic detergent with a hydrophilic maltose head and a hydrophobic long chain alkyl tail. It is considered a gentle detergent that is more efficient than other detergents. For functional solubilization and purification of membrane proteins. It helps to retain the native conformation and activity of membrane-associated proteins and facilitates the reforming of these proteins after denaturation. For the stabilization and activation of enzymes. The critical micelle concentration of DDM is approx. 0.18 mM in water, decreases in the presence of sodium chloride or sucrose and increases in urea.
Catalog No. 50-256-2707