Learn More
Medchemexpress LLC N-ethyl 4-[(3S)-3-methyl-4-morpholinyl]-1H-pyrrolo[2,3-b]pyridine-2-carboxamide | 2109805-96-9 | 99.6% | 288.34 g·mol-1 | C15H20N4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1120815MG
Encompass_Preferred
A substrate-competitive, potent, and selective small-molecule inhibitor of MTH1 (NUDT1) used as a chemical probe for biochemical and cellular studies. Reported IC50 2.3 nM. Molecular formula C15H20N4O2; molecular weight 288.34 g·mol-1.
  • Inhibits MTH1 with high potency (IC50 2.3 nM).
  • Selective scaffold suitable for target validation studies.
  • Demonstrates favorable pharmacokinetic properties in preclinical models.
  • Supplied with supporting analytical data and purity information.
  • Suitable for biochemical assays and cellular target engagement studies.
Catalog No. 50-002-64226