Learn More
Medchemexpress LLC N-ethylglycine | 627-01-0 | MFCD00037794 | 98.0% | 103.12 | C4H9NO2 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW141879250MG
Encompass_Preferred
N-ethylglycine is an N-alkylated glycine derivative provided as a solid for research use. The compound has an analytical purity of 98.0% and a molecular weight of 103.12 g/mol. It is used as an amino acid derivative in biochemical and pharmacological studies, is soluble in DMSO (3.33 mg/mL with ultrasonic and warming), and has defined storage recommendations for powder and solutions.
Catalog No. 50-002-83793