Learn More
Medchemexpress LLC N-Ethylmaleimide | 128-53-0 | 99.89% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD08435MG
Encompass_Preferred
N-Ethylmaleimide (NEM) is a compound derived from maleic acid that can alkylate free sulfhydryl groups. It acts as an irreversible cysteine protease inhibitor and specifically inhibits phosphate transport in mitochondria. It can be used to modify cysteine residues in proteins and peptides.
  • Acts as an irreversible cysteine protease inhibitor
  • Inhibits phosphate transport in mitochondria
  • Modifies cysteine residues in proteins and peptides
  • Inhibits prolyl endopeptidase with an IC50 value of 6.3 μM
Catalog No. 50-004-10631