Learn More
TARGETMOL CHEMICALS INC N-Ethylmaleimide 500MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3088500MG
Encompass_Preferred
Also available in 1 g, 1 mL, 10 mg, 25 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. N-Ethylmaleimide (NEM) is a reagent for alkylation of free sulfhydryl groups, a cysteine protease inhibitor used in experimental biochemistry. N-Ethylmaleimide is also a deubiquitinating enzyme inhibitor that specifically inhibits phosphate transport in mitochondria. Purity 99.92%
Catalog No. 50-287-6311