Learn More
Apexbio Technology LLC N-Ethylmaleimide (NEM) 128-53-0 25mg
SDP
Supplier:  Apexbio Technology LLC C645425
Encompass_Preferred
N-Ethylmaleimide (CAS 128-53-0) is a small molecule commonly utilized in biochemical research as a selective alkylating agent for sulfhydryl groups on cysteine residues in proteins By irreversibly modifying thiol groups N-Ethylmaleimide disrupts the function of enzymes and other proteins that depend on free cysteines for activity or structural integrity This property enables researchers to investigate thiol-dependent biochemical pathways study protein conformational changes and elucidate redox regulation mechanisms Its application extends to proteomics enzymology and the analysis of intracellular signaling processes involving cysteine-mediated modifications
Catalog No. 50-000-89676