Learn More
Selleck Chemical LLC N-Ethylmaleimide (NEM) 25mg 128-53-0
SDP
Supplier:  Selleck Chemical LLC S369225MG
Encompass_Preferred
N-Ethylmaleimide (NEM) is an organic compound that is derived from maleic acid. It is an irreversible inhibitor of all cysteine peptidases, with alkylation occurring at the active site thiol group. N-Ethylmaleimide (NEM) inactivates endogenous deubiquitinating enzymes (DUBs). N-Ethylmaleimide (NEM) specifically inhibits phosphate transport in mitochondria. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8052