Learn More
Medchemexpress LLC N-(fluoren-9-ylmethoxycarbonyl)-L-leucine | 35661-60-0 | MFCD00037133 | 99.9% | 353.42 g/mol | C21H23NO4 | 100 G
SDP
Supplier:  Medchemexpress LLC HY101064100G
Encompass_Preferred
Fmoc-leucine is the fluorenylmethyloxycarbonyl (Fmoc)-protected derivative of L-leucine used as a building block in Fmoc solid-phase peptide synthesis and as a research reagent in peptide chemistry. It is supplied as a high-purity solid suitable for peptide assembly, analytical methods, and other biochemical applications.
  • Used as a protected amino acid in Fmoc solid-phase peptide synthesis.
  • High chemical purity appropriate for research and analytical use.
  • Solid powder form with defined storage recommendations for stability.
  • Available in multiple pack sizes for laboratory scale needs.
Catalog No. 50-002-13626