Learn More
Matrix Scientific N-Fluoro-N'-chloromethyltriethylenediaminebis(tetrafluoroborate), 140681-55-6, MFCD00142607, 25g
Supplier: Matrix Scientific 00285025G
Molecular Formula C7H14B2ClF9N2, Purity 95+%, Molecular Weight 354.26, Melting Point 170°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.