Learn More
STA PHARMACEUTICAL US LLC N-Fmoc-(2S,3S)-3-Amino-2-hydroxy-3-m-tolyl-propionic acid | 1 g | CAS 959581-14-7 | MDL MFCD07363621
Supplier: STA PHARMACEUTICAL US LLC STAAA103581GM
N-Fmoc-(2S,3S)-3-Amino-2-hydroxy-3-m-tolyl-propionic acid is a Amino Acid reagent (Subcategory: Unusual AA) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 959581-14-7
- MDL: MFCD07363621
- InChIKey: YAHIXHKNZKCSQJ-GOTSBHOMSA-N
- Molecular Weight: 417.461
- Molecular Formula: C25H23NO5
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (2S,3S)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-hydroxy-3-(m-tolyl)propanoic acid
- SMILES: CC1=CC([C@@H]([C@@H](C(O)=O)O)NC(OCC2C3=CC=CC=C3C4=CC=CC=C42)=O)=CC=C1
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.