Learn More
STA PHARMACEUTICAL US LLC N-Fmoc-4-bromo-L-tryptophan | 100 g | CAS 2244532-69-0 | InChIKey OUMZDLOCSOPQHB-QHCPKHFHSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAA11610100GM
N-Fmoc-4-bromo-L-tryptophan is a Amino Acid reagent (Subcategory: Trp) sold by WuXi TIDES. Offered in 100 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 2244532-69-0
- MDL: No data
- InChIKey: OUMZDLOCSOPQHB-QHCPKHFHSA-N
- Molecular Weight: 505.368
- Molecular Formula: C26H21BrN2O4
- Purity: ≥95%
- Container Type: 500 mL HDPE
- Pack Size: 100 g
- Net Weight: 100 g
- Gross Weight: 160.5 g
- Commodity Code: 29339980
- Country Of Origin: China
- IUPAC: (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-bromo-1H-indol-3-yl)propanoic acid
- SMILES: O=C(OCC1C2=CC=CC=C2C3=CC=CC=C31)N[C@H](C(O)=O)CC4=CNC5=CC=CC(Br)=C45
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.