Learn More
Medchemexpress LLC N-Fmoc-8-aminooctanoic acid | 126631-93-4 | 98.95% | 381.46 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00903010G
Encompass_Preferred
N-Fmoc-8-aminooctanoic acid functions as a PROTAC linker in the synthesis of PROTACs. It consists of an alkane chain featuring terminal Fmoc-protected amine and carboxylic acid groups. The Fmoc group can be deprotected under basic conditions to yield a free amine, suitable for subsequent conjugations. Additionally, the terminal carboxylic acid can react with primary amine groups in the presence of activators like EDC or HATU to form a stable amide bond.
  • Used as a PROTAC linker in the synthesis of PROTACs.
  • Comprises an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups.
  • Fmoc group is deprotectable under basic conditions to obtain a free amine for further conjugations.
  • Terminal carboxylic acid can react with primary amine groups with activators to form a stable amide bond.
Catalog No. 50-004-53141