Learn More
Medchemexpress LLC Nα-Fmoc-D-tryptophan | 86123-11-7 | 100.0% | 426.46 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00896310MM/1ML
Encompass_Preferred
Fmoc-D-Trp-OH (Nα-Fmoc-D-tryptophan) is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is for research use only and not sold to patients.
  • Used as a biochemical reagent
  • Suitable for life science research
  • Available as a solid and solvent solution
  • Store at -20°C for 3 years (powder) or -80°C for 6 months (in solvent)
  • White to off-white appearance
Catalog No. 50-004-53169