Learn More
STA PHARMACEUTICAL US LLC WuXi TIDES N-Fmoc-ethylenediamine HCl | 391624-46-7, 10GR
Supplier:  STA PHARMACEUTICAL US LLC STAAA1142010GM
Encompass
WuXi TIDES N-Fmoc-ethylenediamine HCl, 391624-46-7 | (9H-fluoren-9-yl)methyl (2-aminoethyl)carbamate hydrochloride | NCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O.Cl | M.F. C17H19ClN2O2, M.W. 318.8
Catalog No. 50-234-4655