Learn More
STA PHARMACEUTICAL US LLC N-Fmoc-L-cysteine methyl ester | 1 g | CAS 233266-69-8 | MDL MFCD30533786
Supplier: STA PHARMACEUTICAL US LLC STAAA122221GM
N-Fmoc-L-cysteine methyl ester is a Amino Acid reagent (Subcategory: Cys) sold by WuXi TIDES. Offered in 1 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 233266-69-8
- MDL: MFCD30533786
- InChIKey: KZRRJSFOJTUXHR-KRWDZBQOSA-N
- Molecular Weight: 357.424
- Molecular Formula: C19H19NO4S
- Purity: ≥95%
- Container Type: 15 mL HDPE
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 7.8 g
- Commodity Code: 29309016
- Country Of Origin: China
- IUPAC: methyl (((9H-fluoren-9-yl)methoxy)carbonyl)-L-cysteinate
- SMILES: COC([C@@H](NC(OCC1C2=CC=CC=C2C3=C1C=CC=C3)=O)CS)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.