Learn More
STA PHARMACEUTICAL US LLC N-Fmoc-N-(3-buten-1-yl)-L-leucine | 25 g | CAS 1188391-88-9 | InChIKey PHBUOMPDTGHXAT-QHCPKHFHSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAA1100625GM
N-Fmoc-N-(3-buten-1-yl)-L-leucine is a Amino Acid reagent (Subcategory: Leu) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 1188391-88-9
- MDL: No data
- InChIKey: PHBUOMPDTGHXAT-QHCPKHFHSA-N
- Molecular Weight: 407.51
- Molecular Formula: C25H29NO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: N-(((9H-fluoren-9-yl)methoxy)carbonyl)-N-(but-3-en-1-yl)-L-leucine
- SMILES: C=CCCN([C@H](C(O)=O)CC(C)C)C(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.