Learn More
Medchemexpress LLC N-Fmoc-N-methoxy-3-aminopropionic acid | 247021-90-5 | 341.36 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0085755G
Encompass_Preferred
N-Fmoc-N-methoxy-3-aminopropionic acid is a Weinreb amide linker and protein cross-linking agent. This product is for research use only.
  • Functions as a Weinreb amide linker
  • Acts as a protein cross-linking agent
Catalog No. 50-004-52716