Learn More
Medchemexpress LLC N-Formyl-Met-Ala-Ser | 17351-32-5 | 98.1% | 335.38 | 1 MG

Supplier: Medchemexpress LLC HYP17561MG
N-Formyl-Met-Ala-Ser is a peptide that binds to formyl peptide receptors on neutrophils. This product is for research use only and not sold to patients.
- Purity: 98.08%
- Molecular weight: 335.38
- Formula: C12H21N3O6S
- Appearance: Solid
- Color: White to off-white
- Sequence: Foemyl-Met-Ala-Ser
- Storage: Sealed, away from moisture and light. Powder: -80°C for 2 years, -20°C for 1 year. In solvent: -80°C for 6 months, -20°C for 1 month
- Solubility: DMSO: 100 mg/mL (298.17 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.