Learn More
Cayman Chemical N-Formyl-Met-Leu-Phe 50mg
SDP
Supplier:  Cayman Chemical 2149550 MG
Encompass_Preferred
A peptide agonist of FPRs; induces cellular activation in phagocytes; induces contractions in isolated guinea pig jejunum (EC50 = 11 nM), proximal colon (EC50 = 3.5 nM), and distal colon (EC50 = 2.2 nM)
Catalog No. 50-298-7386