Learn More
Medchemexpress LLC N-Formyl-Met-Leu-Phe | 59880-97-6 | 99.9% | 437.55 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP022410MG
Encompass_Preferred
N-Formyl-Met-Leu-Phe is a chemotactic peptide and specific ligand of N-formyl peptide receptor (FPR). It is reported to inhibit TNF-alpha secretion, promote osteoblastic commitment, and stimulate osteogenesis through various signaling pathways, making it suitable for diverse biological studies.
  • Specific ligand of N-formyl peptide receptor (FPR)
  • Inhibits TNF-alpha secretion
  • Promotes osteoblastic commitment
  • Suppresses adipogenic commitment
  • Stimulates osteogenesis
  • Induces proinflammatory cytokine gene expression
  • Promotes bone formation
  • Mediates release of calprotectin from PMN
Catalog No. 50-004-26721