Learn More
Medchemexpress LLC N-Formyl-Met-Leu-Phe | 59880-97-6 | 99.9% | 437.55 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP0224
Encompass_Preferred
N-Formyl-Met-Leu-Phe (fMLP) is a chemotactic peptide and a specific ligand of N-formyl peptide receptor (FPR), reported to inhibit TNF-alpha secretion. It acts as a leukocyte chemoattractant, directing the migration of immune effector cells to inflammation sites. This product is for research use only.
  • Chemotactic peptide and specific ligand of N-formyl peptide receptor (FPR)
  • Inhibits TNF-alpha secretion
  • Promotes osteoblastic commitment and suppresses adipogenic commitment
  • Stimulates osteogenesis, increasing osteogenic markers and mineralization
  • Induces proinflammatory cytokine gene expression in human peripheral blood monocytes
  • Promotes bone formation
  • Induces release of calprotectin from polymorphonuclear cells (PMN)
Catalog No. 50-004-26850