Learn More
Medchemexpress LLC N-Formyl-Met-Leu-Phe | 59880-97-6 | 99.9% | 437.55 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHYP0224
Encompass_Preferred
N-Formyl-Met-Leu-Phe (fMLP) is a chemotactic peptide and specific ligand for the N-formyl peptide receptor (FPR). It inhibits TNF-alpha secretion and is used in research on osteogenesis, inflammation, and immune responses.
  • Specific ligand for N-formyl peptide receptor (FPR)
  • Inhibits TNF-alpha secretion
  • Promotes osteoblastic commitment and osteogenesis
  • Suppresses adipogenic commitment
  • Induces proinflammatory cytokine gene expression
  • Synergistically induces inflammatory responses with LPS
  • Promotes bone formation in zebrafish and rabbits
  • Induces calprotectin release from polymorphonuclear cells (PMN)
Catalog No. 50-003-93403